missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Aspartic acid dimethyl ester hydrochloride, 98%, Thermo Scientific Chemicals
$71.96 - $270.27
Chemical Identifiers
CAS | 69630-50-8 |
---|---|
Molecular Formula | C6H12ClNO4 |
Molecular Weight (g/mol) | 197.615 |
MDL Number | MFCD00070384 |
InChI Key | PNLXWGDXZOYUKB-PGMHMLKASA-N |
Synonym | d-aspartic acid dimethyl ester hydrochloride, h-d-asp ome-ome.hcl, h-d-asp ome-oh hcl, r-dimethyl 2-aminosuccinate hydrochloride, d-aspartic acid, dimethyl ester, hydrochloride, d-aspartic acid-1,4-dimethyl ester hydrochloride, 1,4-dimethyl 2r-2-aminobutanedioate hydrochloride, h-d-asp diome . hcl, h-d-asp ome-ome hcl, d-aspartic acid dimethyl ester hcl |
PubChem CID | 51051594 |
IUPAC Name | dimethyl (2R)-2-aminobutanedioate;hydrochloride |
SMILES | COC(=O)CC(C(=O)OC)N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6319703
|
Thermo Scientific Chemicals
H6319703 |
1 g |
Each for $71.96
|
|
||||||
AAH6319706
|
Thermo Scientific Chemicals
H6319706 |
5 g |
Each for $270.27
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69630-50-8 | |
197.615 | |
PNLXWGDXZOYUKB-PGMHMLKASA-N | |
51051594 | |
COC(=O)CC(C(=O)OC)N.Cl |
C6H12ClNO4 | |
MFCD00070384 | |
d-aspartic acid dimethyl ester hydrochloride, h-d-asp ome-ome.hcl, h-d-asp ome-oh hcl, r-dimethyl 2-aminosuccinate hydrochloride, d-aspartic acid, dimethyl ester, hydrochloride, d-aspartic acid-1,4-dimethyl ester hydrochloride, 1,4-dimethyl 2r-2-aminobutanedioate hydrochloride, h-d-asp diome . hcl, h-d-asp ome-ome hcl, d-aspartic acid dimethyl ester hcl | |
dimethyl (2R)-2-aminobutanedioate;hydrochloride |
Specifications
114°C to 119°C | |
C6H12ClNO4 | |
d-aspartic acid dimethyl ester hydrochloride, h-d-asp ome-ome.hcl, h-d-asp ome-oh hcl, r-dimethyl 2-aminosuccinate hydrochloride, d-aspartic acid, dimethyl ester, hydrochloride, d-aspartic acid-1,4-dimethyl ester hydrochloride, 1,4-dimethyl 2r-2-aminobutanedioate hydrochloride, h-d-asp diome . hcl, h-d-asp ome-ome hcl, d-aspartic acid dimethyl ester hcl | |
COC(=O)CC(C(=O)OC)N.Cl | |
1 g | |
51051594 | |
98% | |
D-Aspartic acid dimethyl ester hydrochloride |
69630-50-8 | |
MFCD00070384 | |
PNLXWGDXZOYUKB-PGMHMLKASA-N | |
dimethyl (2R)-2-aminobutanedioate;hydrochloride | |
197.615 | |
197.62 | |
Solid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only