missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Gluconic Acid, 50% Aqueous Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor GL10520LT

 View more versions of this product

Catalog No. 18610159


Explore available promotions

Description

Description

Specifications

Specifications

526-95-4,7732-18-5
Reagent
20 L
Poly Drum

Chemical Identifiers

526-95-4
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.