missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Gluconic Acid, 50% Aqueous Solution, Spectrum™ Chemical
$228.76 - $9101.29
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18610156
|
Spectrum Chemical Mfg Cor
GL1051LT |
1 L | Amber Glass Bottle |
Each for $228.76
|
|
||||
18610158
|
Spectrum Chemical Mfg Cor
GL1054LT |
4 L | Amber Glass Bottle |
Each for $534.53
|
|
||||
18610159
|
Spectrum Chemical Mfg Cor
GL10520LT |
20 L | Poly Drum |
Each for $1,653.44
|
|
||||
18610157
|
Spectrum Chemical Mfg Cor
GL105200LTBL |
200 L | Drum |
Each for $9,101.29
|
|
||||
Chemical Identifiers
526-95-4 |
Specifications
526-95-4,7732-18-5 | |
Reagent | |
1 L | |
Amber Glass Bottle |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.