missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Glucuronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 309880250

 View more versions of this product

Catalog No. AC309880250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D-Glucuronic acid
6556-12-3
White
98%
C6H10O7
25 g
+ 36.50 (20.00°C c=6,H2O,after 2 hrs)
2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
C(=O)C(C(C(C(C(=O)O)O)O)O)O
194.14
CHEBI:47953
98%
98%
158.0°C to 162.0°C
Authentic
Plastic bottle
MFCD00064368
15,4501
+ 36.50
IAJILQKETJEXLJ-QTBDOELSSA-N
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
65041
194.14
Crystalline Powder

Chemical Identifiers

6556-12-3
194.14
IAJILQKETJEXLJ-QTBDOELSSA-N
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
C6H10O7
MFCD00064368
CHEBI:47953
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 229-486-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only