missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Glucuronic acid, 98%, Thermo Scientific Chemicals

$165.42 - $302.67

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
MDL Number MFCD00064368
InChI Key IAJILQKETJEXLJ-QTBDOELSSA-N
ChEBI CHEBI:47953
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC309880100
SDS View Documents
Thermo Scientific Chemicals
309880100
10 g Plastic bottle
Each for $165.42
 
AC309880250
SDS View Documents
Thermo Scientific Chemicals
309880250
25 g Plastic bottle
Each for $302.67
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6556-12-3
194.14
IAJILQKETJEXLJ-QTBDOELSSA-N
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
C6H10O7
MFCD00064368
CHEBI:47953

Specifications

6556-12-3
White
98%
C6H10O7
10 g
+ 36.50 (20.00°C c=6,H2O,after 2 hrs)
2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
C(=O)C(C(C(C(C(=O)O)O)O)O)O
194.14
CHEBI:47953
98%
D-Glucuronic acid, 98%
158.0°C to 162.0°C
Authentic
Plastic bottle
MFCD00064368
15,4501
+ 36.50
IAJILQKETJEXLJ-QTBDOELSSA-N
(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
65041
194.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 229-486-4

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only