missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 204571000

 View more versions of this product

Catalog No. AC204571000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D(+)-Glucuronic acid, sodium salt, monohydrate
134°C to 140°C
Authentic
Plastic bottle
MFCD00151051
03, 886
+21° (20°C c=2,H2O,after 4hrs.)
sodium d-glucuronate
C(=O)C(C(C(C(C(=O)O)O)O)O)O.[Na]
234.14
234.14
Crystalline Powder
14984-34-0
White
99%
C6H9NaO7·H2O
100 g
10, 4327
+ 21
QKHMTHNLNZGTSP-JSCKKFHOSA-N
sodium;(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
87235405
99%

Chemical Identifiers

14984-34-0
234.14
QKHMTHNLNZGTSP-JSCKKFHOSA-N
87235405
C(=O)C(C(C(C(C(=O)O)O)O)O)O.[Na]
C6H9NaO7·H2O
MFCD00151051
sodium d-glucuronate
sodium;(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 239-065-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only