Learn More
D-Lactic acid, Thermo Scientific Chemicals
Useful chiral synthon, building block for depsipeptides
$474.81 - $1513.90
Chemical Identifiers
CAS | 10326-41-7 |
---|---|
Molecular Formula | C3H6O3 |
Molecular Weight (g/mol) | 90.078 |
MDL Number | MFCD00068313 |
InChI Key | JVTAAEKCZFNVCJ-UWTATZPHSA-N |
Synonym | d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure |
PubChem CID | 61503 |
ChEBI | CHEBI:42111 |
IUPAC Name | (2R)-2-hydroxypropanoic acid |
SMILES | CC(C(=O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ61871MD
|
Thermo Scientific Chemicals
J61871MD |
250 mg |
Each for $474.81
|
|
|||||
AAJ6187103
|
Thermo Scientific Chemicals
J6187103 |
1 g |
Each for $1,513.90
|
|
|||||
Description
D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.
Solubility
Soluble in water (50mg/ml)
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents, strong bases.
Chemical Identifiers
10326-41-7 | |
90.078 | |
JVTAAEKCZFNVCJ-UWTATZPHSA-N | |
61503 | |
(2R)-2-hydroxypropanoic acid |
C3H6O3 | |
MFCD00068313 | |
d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure | |
CHEBI:42111 | |
CC(C(=O)O)O |
Specifications
10326-41-7 | |
C3H6O3 | |
1720254 | |
d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure | |
JVTAAEKCZFNVCJ-UWTATZPHSA-N | |
(2R)-2-hydroxypropanoic acid | |
61503 | |
90.08 | |
−0.4° (c=8 in Water) | |
D-Lactic acid |
White | |
MFCD00068313 | |
14,5335 | |
Soluble in water (50mg/ml) | |
CC(C(=O)O)O | |
90.078 | |
CHEBI:42111 | |
250 mg | |
Low Melting Solid |
Safety and Handling
GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.
P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
H315-H318
EINECSNumber : 233-713-2
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only