missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Malic acid, 98%
CAS: 636-61-3 | C4H6O5 | 134.09 g/mol
$84.48 - $760.53
Chemical Identifiers
| CAS | 636-61-3 |
|---|---|
| Molecular Formula | C4H6O5 |
| Molecular Weight (g/mol) | 134.09 |
| MDL Number | MFCD00004245 |
| InChI Key | BJEPYKJPYRNKOW-UHFFFAOYNA-N |
| Synonym | d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate |
| PubChem CID | 92824 |
| ChEBI | CHEBI:30796 |
| IUPAC Name | (2R)-2-hydroxybutanedioic acid |
| SMILES | OC(CC(O)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC153500010
|
Thermo Scientific Chemicals
153500010 |
1 g | Glass bottle |
Each for $84.48
|
|
||||
|
AC153500250
|
Thermo Scientific Chemicals
153500250 |
25 g | Glass bottle |
Each for $760.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 636-61-3 | |
| 134.09 | |
| BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
| 92824 | |
| (2R)-2-hydroxybutanedioic acid |
| C4H6O5 | |
| MFCD00004245 | |
| d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate | |
| CHEBI:30796 | |
| OC(CC(O)=O)C(O)=O |
Specifications
| 99.0°C to 104.0°C | |
| White | |
| C4H6O5 | |
| MFCD00004245 | |
| 15, 5771 | |
| Solubility in water: soluble. | |
| OC(CC(O)=O)C(O)=O | |
| 134.09 | |
| CHEBI:30796 | |
| 98% | |
| Authentic | |
| Crystalline Powder |
| 636-61-3 | |
| 98% | |
| HO2CCH2CH(OH)CO2H | |
| 03, 417 | |
| d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate | |
| BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
| (2R)-2-hydroxybutanedioic acid | |
| 92824 | |
| 134.09 | |
| 1 g | |
| Glass bottle | |
| D(+)-Malic acid |
Safety and Handling
GHS P Statement Causes serious eye irritation.
GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
RUO – Research Use Only