missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-Phenyllactic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 344580010

 View more versions of this product

Catalog No. AC344580010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

D(+)-Phenyllactic acid
7326-19-4
98%
C9H10O3
MFCD00078062
+18.5° (20°C c=1,CH3OH)
r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-phenyllactic acid, r-phenyllactate, r-phenyllactate, r-+-3-phenyllactic acid, r-3-phenyllactic acid, r-, r-
O[C@H](CC1=CC=CC=C1)C(O)=O
166.18
CHEBI:32978
98%
98%
Authentic
Glass bottle
C6H5CH2CH(OH)COOH
1g
+ 18.5
VOXXWSYKYCBWHO-MRVPVSSYSA-N
(2R)-2-hydroxy-3-phenylpropanoic acid
643327
166.18

Chemical Identifiers

7326-19-4
166.18
VOXXWSYKYCBWHO-MRVPVSSYSA-N
643327
(2R)-2-hydroxy-3-phenylpropanoic acid
C9H10O3
MFCD00078062
r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-phenyllactic acid, r-phenyllactate, r-phenyllactate, r-+-3-phenyllactic acid, r-3-phenyllactic acid, r-, r-
CHEBI:32978
O[C@H](CC1=CC=CC=C1)C(O)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-803-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only