missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-Phenyllactic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 7326-19-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00078062
InChI Key VOXXWSYKYCBWHO-MRVPVSSYSA-N
Synonym r-2-hydroxy-3-phenylpropionic acid, a-hydroxy-3-phenylpropionic acid, 2r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-phenyllactic acid, r-phenyllactate, r-phenyllactate, r-+-3-phenyllactic acid, 3-phenyl-d-lactic acid, r-, r-
PubChem CID 643327
ChEBI CHEBI:32978
IUPAC Name (2R)-2-hydroxy-3-phenylpropanoic acid
SMILES O[C@H](CC1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC344580010
SDS View Documents
Thermo Scientific Chemicals
344580010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC344580050
SDS View Documents
Thermo Scientific Chemicals
344580050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7326-19-4
166.18
VOXXWSYKYCBWHO-MRVPVSSYSA-N
643327
(2R)-2-hydroxy-3-phenylpropanoic acid
C9H10O3
MFCD00078062
r-2-hydroxy-3-phenylpropionic acid, a-hydroxy-3-phenylpropionic acid, 2r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-phenyllactic acid, r-phenyllactate, r-phenyllactate, r-+-3-phenyllactic acid, 3-phenyl-d-lactic acid, r-, r-
CHEBI:32978
O[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

7326-19-4
98%
C9H10O3
MFCD00078062
+18.5° (20°C c=1,CH3OH)
r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-phenyllactic acid, r-phenyllactate, r-phenyllactate, r-+-3-phenyllactic acid, r-3-phenyllactic acid, r-, r-
O[C@H](CC1=CC=CC=C1)C(O)=O
166.18
CHEBI:32978
98%
Authentic
Glass bottle
C6H5CH2CH(OH)COOH
1g
+ 18.5
VOXXWSYKYCBWHO-MRVPVSSYSA-N
(2R)-2-hydroxy-3-phenylpropanoic acid
643327
166.18
D(+)-Phenyllactic acid, 98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-803-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only