missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Proline methyl ester hydrochloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 445590050

 View more versions of this product

Catalog No. AC445590050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

D-Proline methyl ester hydrochloride
Authentic
Glass Bottle
5g
d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride
COC(=O)C1CCCN1.Cl
165.62
165.62
65365-28-8
97.5 to max. 102.5% min. (Argentometry)
C6H11NO2·ClH
35
HQEIPVHJHZTMDP-NUBCRITNSA-N
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
44629857
98%

Chemical Identifiers

65365-28-8
165.62
d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
C6H11NO2·ClH
HQEIPVHJHZTMDP-NUBCRITNSA-N
44629857
COC(=O)C1CCCN1.Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only