missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Proline methyl ester hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 65365-28-8 |
---|---|
Molecular Formula | C6H11NO2·ClH |
Molecular Weight (g/mol) | 165.62 |
InChI Key | HQEIPVHJHZTMDP-NUBCRITNSA-N |
Synonym | d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride |
PubChem CID | 44629857 |
IUPAC Name | methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
SMILES | COC(=O)C1CCCN1.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC445590050
|
Thermo Scientific Chemicals
445590050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
65365-28-8 | |
165.62 | |
d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride | |
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
C6H11NO2·ClH | |
HQEIPVHJHZTMDP-NUBCRITNSA-N | |
44629857 | |
COC(=O)C1CCCN1.Cl |
Specifications
65365-28-8 | |
97.5 to max. 102.5% min. (Argentometry) | |
C6H11NO2·ClH | |
35 | |
HQEIPVHJHZTMDP-NUBCRITNSA-N | |
methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride | |
44629857 | |
98% |
Authentic | |
Glass Bottle | |
5g | |
d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride | |
COC(=O)C1CCCN1.Cl | |
165.62 | |
165.62 | |
D-Proline methyl ester hydrochloride |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only