missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Serine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals
$51.14 - $531.42
Chemical Identifiers
CAS | 5874-57-7 |
---|---|
Molecular Formula | C23H19Br2N3O3S |
Molecular Weight (g/mol) | 577.29 |
MDL Number | MFCD00066121 |
InChI Key | LYRWWUCOZBTOPL-UHFFFAOYNA-N |
Synonym | d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci |
PubChem CID | 11446470 |
SMILES | CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6368506
|
Thermo Scientific Chemicals
H6368506 |
5 g |
Each for $51.14
|
|
||||||
AAH6368514
|
Thermo Scientific Chemicals
H6368514 |
25 g |
Each for $157.22
|
|
||||||
AAH6368522
|
Thermo Scientific Chemicals
H6368522 |
100 g |
Each for $531.42
|
|
||||||
Description
D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.
Solubility
Soluble in ethanol and methanol.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
5874-57-7 | |
577.29 | |
LYRWWUCOZBTOPL-UHFFFAOYNA-N | |
11446470 |
C23H19Br2N3O3S | |
MFCD00066121 | |
d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci | |
CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1 |
Specifications
163°C to 166°C (decomposition) | |
C23H19Br2N3O3S | |
d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci | |
LYRWWUCOZBTOPL-UHFFFAOYNA-N | |
methyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride | |
577.29 | |
155.58 | |
Solid |
5874-57-7 | |
MFCD00066121 | |
Soluble in ethanol and methanol. | |
CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1 | |
5 g | |
11446470 | |
95% | |
D-Serine methyl ester hydrochloride |
Safety and Handling
P280-P305+P351+P338-P310
H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only