missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Tartaric acid, 99%

CAS: 147-71-7 | C4H6O6 | 150.09 g/mol

$85.37 - $1846.35

Chemical Identifiers

CAS 147-71-7
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00004238
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
PubChem CID 439655
ChEBI CHEBI:15672
SMILES OC(C(O)C(O)=O)C(O)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC137860250
SDS View Documents
Thermo Scientific Chemicals
137860250
25 g Glass Bottle
Each for $85.37
Only null left
 
AC137861000
SDS View Documents
Thermo Scientific Chemicals
137861000
100 g Plastic bottle
Each for $241.34
Only null left
 
AC137862500
SDS View Documents
Thermo Scientific Chemicals
137862500
250 g Plastic bottle
Each for $461.02
Only null left
 
AC137860010
SDS View Documents
Thermo Scientific Chemicals
137860010
1 kg Plastic bottle
Each for $1,846.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

147-71-7
150.09
FEWJPZIEWOKRBE-UHFFFAOYNA-N
439655
OC(C(O)C(O)=O)C(O)=O
C4H6O6
MFCD00004238
d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
CHEBI:15672

Specifications

167°C to 171°C
White
C4H6O6
MFCD00004238
16,304
d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
OC(C(O)C(O)=O)C(O)=O
150.09
CHEBI:15672
≥98.5%
Authentic
Glass Bottle
D(-)-Tartaric acid, 99%
147-71-7
99%
HO2CCH(OH)CH(OH)CO2H
03, 520
15, 9203
FEWJPZIEWOKRBE-UHFFFAOYNA-N
(2S,3S)-2,3-dihydroxybutanedioic acid
439655
150.09
25 g
0.5% max. (1 g, 105°C, 3 hrs)
Crystals or Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-695-6

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only