missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Daidzein, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 328230250

 View more versions of this product

Catalog No. AC328230250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Daidzein
White
98%
C15H10O4
25 mg
15, 2798
Solubility in water: insoluble. Other solubilities: soluble in dmso
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
254.24
CHEBI:28197
98%
486-66-8
Authentic
Glass bottle
MFCD00016954
18,II,1
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
5281708
254.24
Powder

Chemical Identifiers

486-66-8
254.24
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
5281708
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
C15H10O4
MFCD00016954
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
CHEBI:28197
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 207-635-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only