missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Daidzein, 98%, Thermo Scientific Chemicals

$178.02 - $583.86

Chemical Identifiers

CAS 486-66-8
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00016954
InChI Key ZQSIJRDFPHDXIC-UHFFFAOYSA-N
Synonym daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
PubChem CID 5281708
ChEBI CHEBI:28197
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC328230250
SDS View Documents
Thermo Scientific Chemicals
328230250
25 mg Glass bottle
Each for $178.02
 
AC328231000
SDS View Documents
Thermo Scientific Chemicals
328231000
100 mg Glass bottle
Each for $583.86
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

486-66-8
254.24
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
5281708
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
C15H10O4
MFCD00016954
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
CHEBI:28197
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O

Specifications

486-66-8
Authentic
Glass bottle
MFCD00016954
18,II,1
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one
5281708
254.24
Powder
White
98%
C15H10O4
25 mg
15, 2798
Solubility in water: insoluble. Other solubilities: soluble in dmso
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O
254.24
CHEBI:28197
98%
Daidzein
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 207-635-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only