Learn More
Daidzein, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 328231000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Daidzein | |
White | |
98% | |
C15H10O4 | |
100 mg | |
15, 2798 | |
Solubility in water: insoluble. Other solubilities: soluble in dmso | |
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O | |
254.24 | |
CHEBI:28197 | |
98% |
486-66-8 | |
Authentic | |
Glass bottle | |
MFCD00016954 | |
18,II,1 | |
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone | |
ZQSIJRDFPHDXIC-UHFFFAOYSA-N | |
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one | |
5281708 | |
254.24 | |
Powder |
Chemical Identifiers
486-66-8 | |
254.24 | |
ZQSIJRDFPHDXIC-UHFFFAOYSA-N | |
5281708 | |
7-hydroxy-3-(4-hydroxyphenyl)chromen-4-one |
C15H10O4 | |
MFCD00016954 | |
daidzein, 4',7-dihydroxyisoflavone, daidzeol, 7,4'-dihydroxyisoflavone, 7-hydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, diadzein, 7-hydroxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 7-hydroxy-3-4-hydroxyphenyl, 7-hydroxy-3-4-hydroxyphenyl chromen-4-one, 7-hydroxy-3-4-hydroxyphenyl-4-benzopyrone | |
CHEBI:28197 | |
C1=CC(=CC=C1C2=COC3=C(C2=O)C=CC(=C3)O)O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 207-635-4
RUO – Research Use Only