Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
Supplier: Thermo Scientific™ 195898
Description
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:- RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
- RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
Specifications
RS-485; 0-1V | |
Revco™ CxF, HERAfreeze™ HFC, Model 1535 Monitor/Alarm System, TSC Series | |
0/1 V |
Output | |
Alarm, Recorder |