Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
$1193.45 - $1512.43
Specifications
Monitoring Options Alarm, Recorder
View More Specs
Products 7
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
11-676-200
View Documents
Thermo Scientific™
195895
RS-232; 0-1V
Each for $1,204.30
11-676-201
View Documents
Thermo Scientific™
195896
RS-232; 0-5V
Each for $1,228.17
11-676-202
View Documents
Thermo Scientific™
195897
RS-485; 4-20mA
Each for $1,512.43
11-676-203
View Documents
Thermo Scientific™
195898
RS-485; 0-1V
Each for $1,219.49
11-676-204
View Documents
Thermo Scientific™
195899
RS-485; 0-5V
Each for $1,193.45
13-990-503
View Documents
Thermo Scientific™
398213
RS-232, 0-1V (Factory Installed Only)
Each for $1,375.91
13-990-504
View Documents
Thermo Scientific™
398214
RS-485, 0-1V (Factory Installed Only)
Each for $1,393.65
Description
Description
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:
  • RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
  • RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
Specifications
Specifications
Alarm, Recorder
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'