Corning™ DeckWorks™ Reload Systems Starter Kits
Contains one DeckWorks Reload System with standard tips and empty hinged racks
Supplier: Corning™ 4133
Description
DeckWorks starter kits contain one DeckWorks reload system with standard tips and empty hinged racks.
- 10 empty racks accompany 10 μL, 200 μL, and 300 μL volume tips.
- 8 empty racks accompany 1000 μL volume tips.
Specifications
1 to 300 μL | |
Natural | |
Refill | |
Starter kit | |
DeckWorks |
Non-sterile | |
Designed to allow to maximize storage space and help reduce the production and waste of plastic materials | |
Round | |
Yes | |
Polypropylene |