Decon™ Contrad™ 100 Equipment Cleaner
Formulated for clean-in-place systems and high-pressure cleaning applications
Supplier: Decon™ 1506
Description
- Alkaline liquid detergent Removes a variety of contaminants across a wide range of temperatures and pressures
- Low-foaming, free-rinsing, phosphate free, and disposable to drain
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
- Non-toxic: Item is non-toxic
Specifications
Liquid | |
Sterile | |
100°C | |
30 gal. |
12.4 | |
Clear | |
For clean in place systems and high pressure cleaning applications |