missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decon™ Contrad™ 100 Equipment Cleaner

Formulated for clean-in-place systems and high-pressure cleaning applications

$688.70 - $6411.20

Specifications

Form Liquid
pH 12.4
Sterility Sterile
Appearance Clear
Boiling Point 100°C
View More Specs

Products 4
Catalog Number Mfr. No. Volume (English) Price Quantity  
Catalog Number Mfr. No. Volume (English) Price Quantity  
04-355-27
View Documents
Decon™
1504
1 gal.
Case of 4 Each for $688.70
 
04-355-370
View Documents
Decon™
1506
30 gal.
Each for $3,175.23
 
04-355-55
View Documents
Decon™
DC1503
5.25 gal.
Each for $697.03
 
04-355-57
View Documents
Decon™
DC1505
55.5 gal.
Each for $6,411.20
 
Description

Description

  • Alkaline liquid detergent Removes a variety of contaminants across a wide range of temperatures and pressures
  • Low-foaming, free-rinsing, phosphate free, and disposable to drain
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic

Specifications

Specifications

Liquid
Sterile
100°C
12.4
Clear
For clean in place systems and high pressure cleaning applications
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'