Decon™ Contrad™ NF Liquid Detergent
Cleans a wide variety of organic and inorganic contaminants from most substrates
Supplier: Decon™ DC6003
Description
- Super-concentrated alkaline liquid
- Formulated specifically for automatic washing machines
- Nonfoaming, free-rinsing, and residue-free
- Phosphate- and chlorine-free
- Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
- Recycled: Item contains recycled content as described in the item detail
- Non-toxic: Item is non-toxic
Specifications
Liquid | |
>100°C | |
30.33 gal. |
14 | |
For automatic washing machines, removes Organic and Inorganic contaminants |