missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Decon™ Contrad™ NF Liquid Detergent

Cleans a wide variety of organic and inorganic contaminants from most substrates

$198.00 - $3,885.13

Specifications

Form Liquid
pH 14
Boiling Point >100°C
For Use With (Application) For automatic washing machines, removes Organic and Inorganic contaminants
View More Specs

Products 3
Catalog Number Mfr. No. Volume (English) Price Quantity  
Catalog Number Mfr. No. Volume (English) Price Quantity  
04-355-30
View Documents
Decon™
6001
2.62 gal. N/A
 
04-355-37
View Documents
Decon™
DC6002
1 gal. N/A
 
04-355-9
View Documents
Decon™
DC6003
30.33 gal. N/A
 
Description

Description

  • Super-concentrated alkaline liquid
  • Formulated specifically for automatic washing machines
  • Nonfoaming, free-rinsing, and residue-free
  • Phosphate- and chlorine-free
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail
  • Non-toxic: Item is non-toxic

Specifications

Specifications

Liquid
14
>100°C
For automatic washing machines, removes Organic and Inorganic contaminants
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'