Eppendorf™ DNA LoBind™ Plates
DNA LoBind Plates maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.
Supplier:  Eppendorf™ 951031607
 View more versions of this product
Catalog No. 05414221
Explore available promotions
Description
Description
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results.
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing.
Specifications
Specifications
Autoclavable
White
Polypropylene
384
DNA Recovery
PCR Clean (Non-sterile)
Colorless
120 Plates (10 Bags x 12 Plates)
DNA LoBind™ Plate
200 μL
Non-sterile
6000 x g
120 Pk.