missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ DNA LoBind™ Plates
DNA LoBind Plates maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.
$262.63 - $1998.17
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
Max. RCF | 6000 x g |
Purity or Quality Grade | PCR Clean (Non-sterile) |
Catalog Number | Mfr. No. | No. of Wells | Volume (Metric) Well | Color | Packaging | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Wells | Volume (Metric) Well | Color | Packaging | Price | Quantity | ||
E951032000
|
Eppendorf™
951032000 |
96 | 500 μL | White | 40 Plates (5 Bags x 8 Plates) |
Pack of 40 Each for $713.60
|
|
||
05414229
|
Eppendorf™
951032085 |
96 | 500 μL | Blue | 40 Plates (5 Bags x 8 Plates) |
Case of 40 Each for $673.21
|
|
||
E951032409
|
Eppendorf™
951032409 |
96 | 500 μL | White, Clear Bottom | 120 Plates (10 Bags x 12 Plates) |
Pack of 120 Each for $1,811.65
|
|
||
05414238
|
Eppendorf™
951032883 |
96 | 1000 μL | Blue | 20 Plates (5 Bags x 4 Plates) |
Case of 20 Each for $262.63
|
|
||
E951032808
|
Eppendorf™
951032808 |
96 | 1000 μL | White, White | 20 Plates (5 Bags x 4 Plates) |
Pack of 20 Each for $262.63
|
|
||
E951033201
|
Eppendorf™
951033201 |
96 | 1000 μL | White, Clear Bottom | 80 Plates (10 Bags x 8 Plates) |
Pack of 80 Each for $915.48
|
|
||
05414216
|
Eppendorf™
0030603303 |
96 | 350 μL | White, Clear | 80 Plates (5 Bags x 16 Plates) |
Case of 80 Each for $1,329.37
|
|
||
E951040546
|
Eppendorf™
951040546 |
384 | 140 μL | White | 80 Plates (5 Bags x 16 Plates) |
Each for $1,459.11
|
|
||
E951031208
|
Eppendorf™
951031208 |
384 | 200 μL | White | 40 Plates (5 Bags x 8 Plates) |
Pack of 40 Each for $749.32
|
|
||
05414221
|
Eppendorf™
951031607 |
384 | 200 μL | White | 120 Plates (10 Bags x 12 Plates) |
Case of 120 Each for $1,998.17
|
|
||
Description
- Eppendorf LoBind material ensures optimized sample recovery for improved assay results.
- Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
- Raised well rims and a smooth surface ensure reliable sealing.
Specifications
Autoclavable | |
Non-sterile | |
PCR Clean (Non-sterile) |
Polypropylene | |
6000 x g | |
Colorless |