missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eppendorf™ DNA LoBind™ Plates
DNA LoBind Plates maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.
Supplier: Eppendorf™ 951032883
Description
- Eppendorf LoBind material ensures optimized sample recovery for improved assay results.
- Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
- Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
- High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
- Raised well rims and a smooth surface ensure reliable sealing.
Specifications
Autoclavable | |
Blue | |
Polypropylene | |
96 | |
DNA Recovery | |
PCR Clean (Non-sterile) | |
Colorless |
20 Plates (5 Bags x 4 Plates) | |
DNA LoBind™ Plate | |
1000 μL | |
Non-sterile | |
6000 x g | |
20 Pk. |