missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eppendorf™ DNA LoBind™ Plates

DNA LoBind Plates maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.

Supplier:  Eppendorf™ 951040546

 View more versions of this product

Catalog No. E951040546


Explore available promotions

Description

Description

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results.
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference.
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors.
  • High-contrast Unique OptiTrack™ matrix: up to 30% faster sample identification and fewer pipetting errors.
  • RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties.
  • Raised well rims and a smooth surface ensure reliable sealing.
Specifications

Specifications

Standard
Autoclavable
White
Polypropylene
384
Conical Bottom (V-well)
DNA or RNA applications in forensic analysis, microarrays and next generation sequencing
PCR Clean (Non-sterile)
Yes
12.77 cm
8.55 cm
16 x 24
80 Plates (5 Bags x 16 Plates)
384 Deep-Well Microplate
140 μL
Non-sterile
Yes
6000 x g
80 Pk.
Colorless
1.44 cm
Videos
SDS
Documents

Documents

Promotions

Promotions