missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Deluxe Office Monitor Arm

Deluxe monitor arm effortlessly adjusts to the angle and focal depth of your flat screen monitor for either seated or standing applications.

Supplier:  Fisherbrand™ FS103S

 View more versions of this product

Catalog No. 06001038


Explore available promotions

Description

Description

  • 4 to 21 in. horizontal travel range
  • Accommodates all VESA-compliant monitors weighting up to 25 lb.
  • In clamp or grommet mounts
  • Portrait or landscape monitor compatible
Specifications

Specifications

4 in.
10.2 cm
Single Arm
45.7 cm
18 in.
Silver
Videos
SDS
Documents

Documents

Promotions

Promotions