Thermo Scientific™ Dense Storage Options
Double your cryobank capacity and cut storage costs by 50%. Our dense storage options fit standard cryogenic storage boxes that work with the freezer racks you currently own.
$223.91 - $3921.68
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
12565004
|
Thermo Scientific™
374001 |
Cryobox divider, 13 x 13 grid |
Case of 24 Each for $680.62
|
|
|||||
15-350-55
|
Thermo Scientific™
50500001 |
Polycarbonate CryoBox, Undivided |
Pack of 4 Each for $233.97
Case of 6 Pack for $1,403.82
|
|
|||||
15-350-501
|
Thermo Scientific™
50500169 |
Polycarbonate CryoBox with 13 x 13 divider |
Pack of 4 Each for $223.91
Case of 6 Pack for $1,343.46
|
|
|||||
12-566-298
|
Thermo Scientific™
374115 |
1.0mL Nunc Cryobank Tubes, Blank, with Caps, Sterile, Loose Packed |
Case of 845 Each for $3,921.68
|
|
|||||
Description
Double your cryobank storage capacity and cut storage costs by 50%. The Thermo Scientific™ Dense Storage Options use standard CryoBoxes™ with 13 x 13 tube separators and fit 169 1mL tubes in a single CryoBox™. These standard-sized boxes are compatible with existing standard-sized lab freezer racks.
- Choose a standard-sized cardboard or polycarbonate box
- Insert the 13x13 box divider
- Load the box with 169, 1mL Cryobank storage tubes
- Slide the box into your existing standard-sized CryoBox racks
- Suitable for mechanical freezer or vapor phase liquid nitrogen storage