missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 10 to 15%, C13H13IO8, CAS Number-87413-09-0, 75-09-2, dess martin, dess-martin, dess-martinperiodinane, dess-martin periodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess martin periodinane, dess-martin periodane, dess-martin reagent, triacetoxyperiodinane
Supplier: Thermo Scientific Chemicals 333110500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagentSpecifications
Dess-Martin periodinane | |
15 wt. % Solution in Dichloromethane | |
C13H13IO8 | |
1.3620g/mL | |
NKLCNNUWBJBICK-UHFFFAOYSA-N | |
(1,1-diacetyloxy-3-oxo-1$l^{5},2-benziodoxol-1-yl) acetate | |
159087 | |
10 to 15% | |
1.362 | |
Liquid |
Colorless to Yellow | |
87413-09-0,75-09-2 | |
MFCD00130127 | |
dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane | |
CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12 | |
424.14 | |
424.15 | |
Glass bottle | |
50 mL |
RUO – Research Use Only