missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals

Dess-Martin periodinane, 10 to 15%, C13H13IO8, CAS Number-87413-09-0, 75-09-2, dess martin, dess-martin, dess-martinperiodinane, dess-martin periodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess martin periodinane, dess-martin periodane, dess-martin reagent, triacetoxyperiodinane

$96.53 - $450.02

Specifications

Chemical Name or Material Dess-Martin periodinane
CAS 87413-09-0,75-09-2
Molecular Formula C13H13IO8
MDL Number MFCD00130127
Density 1.3620g/mL
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC333110500
View Documents
Thermo Scientific Chemicals
333110500
50 mL
Each for $450.02
 
AC333110100
View Documents
Thermo Scientific Chemicals
333110100
10 mL
Each for $96.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
Specifications

Specifications

Dess-Martin periodinane
C13H13IO8
1.3620g/mL
NKLCNNUWBJBICK-UHFFFAOYSA-N
424.14
424.15
87413-09-0,75-09-2
MFCD00130127
dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12
159087
1.362
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only