missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-n-butylamine, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A11671AP

 View more versions of this product

Catalog No. AAA11671AP


Explore available promotions

Description

Description

Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-butylamine is an important intermediate used for various applications in organic synthesis. It is involved in the production of rubber chemicals like di-n-butylthiocarbamate thiuram, emulsifiers and dibutylethanolamine, which is used in coatings and paints. It is also used as plasticizer in PVC matrix and a corrosion inhibitor in an aqueous system. It plays an important role as a catalyst in vulcanization.

Solubility
Miscible with water, ether, ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, carbon dioxide, zinc, iron and copper.
Specifications

Specifications

Di-n-butylamine
°C to 62°C
159°C to 160°C
≥98%
1.4169
MFCD00009429
UN2248
14,3032
Miscible with water,ether,ethanol and acetone.
CCCCNCCCC
129.247
129.25
Liquid
111-92-2
0.767
39°C (102°F)
C8H19N
[CH3(CH2)3]2NH
500 mL
506001
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
JQVDAXLFBXTEQA-UHFFFAOYSA-N
N-butylbutan-1-amine
8148
≥98%

Chemical Identifiers

111-92-2
129.247
JQVDAXLFBXTEQA-UHFFFAOYSA-N
8148
CCCCNCCCC
C8H19N
MFCD00009429
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
N-butylbutan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H314-H318-H226

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H301+H311-H314-H330

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: DI-N-BUTYLAMINE

EINECSNumber : 203-921-8

RTECSNumber : HR7780000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only