missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-tert-butyl dicarbonate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 189771000

 View more versions of this product

Catalog No. AC189771000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Di-tert-butyl Dicarbonate
24424-99-5
White
56.0°C to 57.0°C (0.5 mmHg)
Authentic
Glass Bottle
1.4075 to 1.4095
MFCD00008805
2930
1.02
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
218.25
CHEBI:48500
97%
97%
22.0°C to 24.0°C
1.0200g/mL
37°C
97%
C10H18O5
O[CO2C(CH3)3]2
100 g
04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113
15, 3038
DYHSDKLCOJIUFX-UHFFFAOYSA-N
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
90495
218.25
Low Melting Crystalline Solid

Chemical Identifiers

24424-99-5
218.25
DYHSDKLCOJIUFX-UHFFFAOYSA-N
90495
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
C10H18O5
MFCD00008805
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
CHEBI:48500
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
Fatal if inhaled.
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cauti

GHS Signal Word: Danger

EINECSNumber : 246-240-1

RTECSNumber : HT0230000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only