missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Di-tert-butyl dicarbonate, 97%, Thermo Scientific Chemicals

$273.23 - $273.23

Chemical Identifiers

CAS 24424-99-5
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.25
MDL Number MFCD00008805
InChI Key DYHSDKLCOJIUFX-UHFFFAOYSA-N
Synonym di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
PubChem CID 90495
ChEBI CHEBI:48500
IUPAC Name tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
SMILES CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC189771000
SDS View Documents
Thermo Scientific Chemicals
189771000
100 g Glass Bottle
Each for $273.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24424-99-5
218.25
DYHSDKLCOJIUFX-UHFFFAOYSA-N
90495
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
C10H18O5
MFCD00008805
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
CHEBI:48500
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

Specifications

24424-99-5
White
56.0°C to 57.0°C (0.5 mmHg)
Authentic
Glass Bottle
1.4075 to 1.4095
MFCD00008805
2930
1.02
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
218.25
CHEBI:48500
97%
Di-tert-butyl Dicarbonate, 97%
22.0°C to 24.0°C
1.0200g/mL
37°C
97%
C10H18O5
O[CO2C(CH3)3]2
100 g
04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113
15, 3038
DYHSDKLCOJIUFX-UHFFFAOYSA-N
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
90495
218.25
Low Melting Crystalline Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause an allergic skin reaction.
Fatal if inhaled.
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cauti

GHS Signal Word: Danger

EINECSNumber : 246-240-1

RTECSNumber : HT0230000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only