Learn More
Di-tert-butyl peroxide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 349932500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsSpecifications
Di-tert-butyl peroxide | |
-30°C | |
109°C to 110°C | |
Authentic | |
Plastic bottle | |
1.3880 to 1.39 | |
MFCD00008803 | |
0.8 | |
di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d | |
LSXWFXONGKSEMY-UHFFFAOYSA-N | |
2-tert-butylperoxy-2-methylpropane | |
8033 | |
146.23 | |
Liquid |
110-05-4 | |
0.8000g/mL | |
6°C | |
0.1% max. Tert-butyl hydroperoxide (GC) | |
C8H18O2 | |
(CH3)3COOC(CH3)3 | |
250 g | |
15, 3508 | |
Solubility in water: immiscible. Other solubilities: soluble in most organic solvents | |
CC(C)(C)OOC(C)(C)C | |
146.23 | |
0.9 mPa.s (20°C) | |
99% |
Chemical Identifiers
110-05-4 | |
146.23 | |
LSXWFXONGKSEMY-UHFFFAOYSA-N | |
8033 |
C8H18O2 | |
MFCD00008803 | |
di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d | |
CC(C)(C)OOC(C)(C)C |
Safety and Handling
GHS H Statement
Heating may cause a fire.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get me
GHS Signal Word: Danger
EINECSNumber : 203-733-6
RUO – Research Use Only