missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl peroxide, 99%

CAS: 110-05-4 | C8H18O2 | 146.23 g/mol

$74.74 - $136.20

Chemical Identifiers

CAS 110-05-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00008803
InChI Key LSXWFXONGKSEMY-UHFFFAOYSA-N
Synonym di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d
PubChem CID 8033
SMILES CC(C)(C)OOC(C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC349930050
SDS View Documents
Thermo Scientific Chemicals
349930050
5 g Plastic bottle
Each for $74.74
Only null left
 
AC349931000
SDS View Documents
Thermo Scientific Chemicals
349931000
100 g Plastic bottle
Each for $82.10
Only null left
 
AC349932500
SDS View Documents
Thermo Scientific Chemicals
349932500
250 g Plastic bottle
Each for $136.20
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
Specifications

Chemical Identifiers

110-05-4
146.23
LSXWFXONGKSEMY-UHFFFAOYSA-N
8033
C8H18O2
MFCD00008803
di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d
CC(C)(C)OOC(C)(C)C

Specifications

110-05-4
0.8000g/mL
6°C
0.1% max. Tert-butyl hydroperoxide (GC)
C8H18O2
(CH3)3COOC(CH3)3
5 g
15, 3508
Solubility in water: immiscible. Other solubilities: soluble in most organic solvents
CC(C)(C)OOC(C)(C)C
146.23
0.9 mPa.s (20°C)
99%
Di-tert-butyl peroxide
-30°C
109°C to 110°C
Authentic
Plastic bottle
1.3880 to 1.39
MFCD00008803
0.8
di-tert-butyl peroxide, tert-butyl peroxide, di-t-butyl peroxide, cadox, peroxide, bis 1,1-dimethylethyl, dtbp, trigonox b, t-butyl peroxide, cadox tbp, kayabutyl d
LSXWFXONGKSEMY-UHFFFAOYSA-N
2-tert-butylperoxy-2-methylpropane
8033
146.23
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Heating may cause a fire.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get me

GHS Signal Word: Danger

EINECSNumber : 203-733-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only