Learn More
Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide, Thermo Scientific Chemicals
Diallyl disulfide, 80%, C6H10S2, CAS Number-2179-57-9, diallyldisulfide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene, 2-propenyl disulphide, diallyl disulfide, allyl disulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 100g
Supplier: Thermo Scientific Chemicals L1110622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diallyl disulfide | |
1.008 | |
51°C (123°F) | |
remainder mainly diallyl sulfide and diallyl trisulfide | |
1.541 | |
MFCD00008656 | |
UN1993 | |
diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene | |
C=CCSSCC=C | |
146.27 | |
CHEBI:4488 | |
80% |
2179-57-9 | |
180°C to 190°C | |
Garlic-like | |
C6H10S2 | |
CH2=CHCH2S-SCH2CH=CH2 | |
100 g | |
1699241 | |
PFRGXCVKLLPLIP-UHFFFAOYSA-N | |
3-(prop-2-enyldisulfanyl)prop-1-ene | |
16590 | |
146.27 | |
Technical |
Chemical Identifiers
2179-57-9 | |
146.27 | |
PFRGXCVKLLPLIP-UHFFFAOYSA-N | |
16590 | |
3-(prop-2-enyldisulfanyl)prop-1-ene |
C6H10S2 | |
MFCD00008656 | |
diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene | |
CHEBI:4488 | |
C=CCSSCC=C |
Safety and Handling
GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P330-P333+P313-P363-P370+P378q-P501c
H226-H301-H315-H317-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 218-548-6
RTECSNumber : BB1000000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only