missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diallyl disulfide, tech. 80%, remainder mainly diallyl sulfide and diallyl trisulfide, Thermo Scientific Chemicals

Diallyl disulfide, 80%, C6H10S2, CAS Number-2179-57-9, diallyldisulfide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene, 2-propenyl disulphide, diallyl disulfide, allyl disulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 100g

$111.58 - $281.23

Chemical Identifiers

CAS 2179-57-9
Molecular Formula C6H10S2
Molecular Weight (g/mol) 146.27
MDL Number MFCD00008656
InChI Key PFRGXCVKLLPLIP-UHFFFAOYSA-N
Synonym diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene
PubChem CID 16590
ChEBI CHEBI:4488
IUPAC Name 3-(prop-2-enyldisulfanyl)prop-1-ene
SMILES C=CCSSCC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1110614
View Documents
Thermo Scientific Chemicals
L1110614
25 g
Each for $111.58
 
AAL1110622
View Documents
Thermo Scientific Chemicals
L1110622
100 g
Each for $281.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2179-57-9
146.27
PFRGXCVKLLPLIP-UHFFFAOYSA-N
16590
3-(prop-2-enyldisulfanyl)prop-1-ene
C6H10S2
MFCD00008656
diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene
CHEBI:4488
C=CCSSCC=C

Specifications

2179-57-9
180°C to 190°C
Garlic-like
1.541
MFCD00008656
UN1993
diallyl disulfide, allyl disulfide, diallyldisulfide, diallyl disulphide, disulfide, di-2-propenyl, 1,2-diallyldisulfane, 2-propenyl disulphide, allyl disulphide, 4,5-dithia-1,7-octadiene, 3-allyldisulfanyl-propene
C=CCSSCC=C
146.27
CHEBI:4488
80%
Diallyl disulfide
1.008
51°C (123°F)
C6H10S2
CH2=CHCH2S-SCH2CH=CH2
25 g
1699241
PFRGXCVKLLPLIP-UHFFFAOYSA-N
3-(prop-2-enyldisulfanyl)prop-1-ene
16590
146.27
Technical
remainder mainly diallyl sulfide and diallyl trisulfide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P330-P333+P313-P363-P370+P378q-P501c

H226-H301-H315-H317-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 218-548-6

RTECSNumber : BB1000000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only