missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diatomaceous Earth, J.T. Baker™
Supplier: Avantor J.T.Baker 193901
Specifications
500 g | |
33 Micron | |
Wide Mouth Amber Glass Bottle | |
CNa2O3 | |
C(=O)([O-])[O-].[Na+].[Na+] | |
105.988 | |
CHEBI:29377 |
1L = 2.30kg | |
Diatomaceous Earth | |
68855-54-9 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 |
Chemical Identifiers
68855-54-9 | |
105.988 | |
10340 | |
disodium;carbonate |
CNa2O3 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
CHEBI:29377 | |
C(=O)([O-])[O-].[Na+].[Na+] |