missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diatomaceous Earth, J.T. Baker™
$4404.44 - $4404.44
Chemical Identifiers
CAS | 68855-54-9 |
---|---|
Molecular Formula | CNa2O3 |
Molecular Weight (g/mol) | 105.988 |
InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
PubChem CID | 10340 |
ChEBI | CHEBI:29377 |
IUPAC Name | disodium;carbonate |
SMILES | C(=O)([O-])[O-].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
02002714
|
Avantor
193901 |
500 g | Wide Mouth Amber Glass Bottle |
Case of 4 Each for $4,404.44
|
|
||||
Chemical Identifiers
68855-54-9 | |
105.988 | |
10340 | |
disodium;carbonate |
CNa2O3 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
CHEBI:29377 | |
C(=O)([O-])[O-].[Na+].[Na+] |
Specifications
500 g | |
33 Micron | |
68855-54-9 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium;carbonate | |
10340 | |
Diatomaceous Earth |
1L = 2.30kg | |
Wide Mouth Amber Glass Bottle | |
CNa2O3 | |
C(=O)([O-])[O-].[Na+].[Na+] | |
105.988 | |
CHEBI:29377 |