missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibenzoylmethane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 112600100

 View more versions of this product

Catalog No. AC112600100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dibenzoylmethane
120-46-7
Yellow
Authentic
Glass Bottle
C6H5COCH2COC6H5
10 g
15,3014
NZZIMKJIVMHWJC-UHFFFAOYSA-N
1,3-diphenylpropane-1,3-dione
8433
224.26
Crystals
98%
75°C to 79°C
219°C to 221°C (18.0 mmHg)
98%
C15H12O2
MFCD00003085
07,769
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2
224.26
CHEBI:75417
98%

Chemical Identifiers

120-46-7
224.26
NZZIMKJIVMHWJC-UHFFFAOYSA-N
8433
1,3-diphenylpropane-1,3-dione
C15H12O2
MFCD00003085
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
CHEBI:75417
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-398-9

RTECSNumber : TZ1930000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only