missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibenzoylmethane, 98%, Thermo Scientific Chemicals

$55.00 - $55.00

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00003085
InChI Key NZZIMKJIVMHWJC-UHFFFAOYSA-N
Synonym dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
PubChem CID 8433
ChEBI CHEBI:75417
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC112600100
SDS View Documents
Thermo Scientific Chemicals
112600100
10 g Glass Bottle
Each for $55.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

120-46-7
224.26
NZZIMKJIVMHWJC-UHFFFAOYSA-N
8433
1,3-diphenylpropane-1,3-dione
C15H12O2
MFCD00003085
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
CHEBI:75417
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2

Specifications

120-46-7
Yellow
Authentic
Glass Bottle
C6H5COCH2COC6H5
10 g
15,3014
NZZIMKJIVMHWJC-UHFFFAOYSA-N
1,3-diphenylpropane-1,3-dione
8433
224.26
Crystals
75°C to 79°C
219°C to 221°C (18.0 mmHg)
98%
C15H12O2
MFCD00003085
07,769
dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2
224.26
CHEBI:75417
98%
Dibenzoylmethane, 98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-398-9

RTECSNumber : TZ1930000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only