missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dibenzyl azodicarboxylate, 94%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 270270050

 View more versions of this product

Catalog No. AC270270050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen), Radical initiators
Specifications

Specifications

Dibenzyl azodicarboxylate
2449-05-0
Yellow to Orange
0.75% max.
94%
C16H14N2O4
5 g
dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate
IRJKSAIGIYODAN-ISLYRVAYSA-N
benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate
5387121
94%
94%
44°C to 52°C
>230°C
Authentic
Glass bottle
MFCD00016737
15,111
Solubility in water: insoluble
C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2
298.29
298.29
Crystalline Powder

Chemical Identifiers

2449-05-0
298.29
IRJKSAIGIYODAN-ISLYRVAYSA-N
5387121
C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2
C16H14N2O4
MFCD00016737
dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate
benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-508-0

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only