Learn More
Dibenzyl azodicarboxylate, 94%
CAS: 2449-05-0 | C16H14N2O4 | 298.29 g/mol
$129.71 - $379.14
Chemical Identifiers
| CAS | 2449-05-0 |
|---|---|
| Molecular Formula | C16H14N2O4 |
| Molecular Weight (g/mol) | 298.29 |
| MDL Number | MFCD00016737 |
| InChI Key | IRJKSAIGIYODAN-ISLYRVAYSA-N |
| Synonym | dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate |
| PubChem CID | 5387121 |
| IUPAC Name | benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate |
| SMILES | C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC270270050
|
Thermo Scientific Chemicals
270270050 |
5 g | Glass bottle |
Each for $129.71
|
|
||||
|
AC270270250
|
Thermo Scientific Chemicals
270270250 |
25 g | Glass bottle |
Each for $379.14
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiatorsChemical Identifiers
| 2449-05-0 | |
| 298.29 | |
| IRJKSAIGIYODAN-ISLYRVAYSA-N | |
| 5387121 | |
| C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2 |
| C16H14N2O4 | |
| MFCD00016737 | |
| dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate | |
| benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate |
Specifications
| 2449-05-0 | |
| Yellow to Orange | |
| 0.75% max. | |
| 94% | |
| C16H14N2O4 | |
| 5 g | |
| dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate | |
| IRJKSAIGIYODAN-ISLYRVAYSA-N | |
| benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate | |
| 5387121 | |
| 94% | |
| Dibenzyl azodicarboxylate, 94% |
| 44°C to 52°C | |
| >230°C | |
| Authentic | |
| Glass bottle | |
| MFCD00016737 | |
| 15,111 | |
| Solubility in water: insoluble | |
| C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2 | |
| 298.29 | |
| 298.29 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 219-508-0
TSCA : TSCA
RUO – Research Use Only