Learn More
Dibutylgermanium dichloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 381110050
Specifications
Dibutylgermanium dichloride | |
Colorless | |
242°C | |
97.5% min. (GC) | |
C8H18Cl2Ge | |
(CH3CH2CH2CH2)2Ge(Cl)2 | |
5 g | |
di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4 | |
CCCC[Ge](CCCC)(Cl)Cl | |
257.73 | |
257.73 | |
Liquid |
4593-81-1 | |
1.2080g/mL | |
Authentic | |
Glass bottle | |
1.4720 to 1.474 | |
MFCD00013590 | |
1.208 | |
VKMPTHGEGBKPBQ-UHFFFAOYSA-N | |
dibutyl(dichloro)germane | |
20723 | |
98% |
Chemical Identifiers
4593-81-1 | |
257.73 | |
VKMPTHGEGBKPBQ-UHFFFAOYSA-N | |
20723 | |
CCCC[Ge](CCCC)(Cl)Cl |
C8H18Cl2Ge | |
MFCD00013590 | |
di-n-butylgermanium dichloride, dibutylgermanium dichloride, dichlorodibutylgermane, germane, dibutyldichloro, di-n-butylgermanedichloride, di-n-butyldichlorogermane, dibutyl dichloro germane, wln: g-ge-g4&4 | |
dibutyl(dichloro)germane |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 224-986-9
RUO – Research Use Only