missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dichlorodimethylgermane, 95+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 316500010

 View more versions of this product

Catalog No. AC316500010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dichlorodimethylgermane
-22°C
1.5000g/mL
32°C
Glass bottle
1.4590 to 1.461
MFCD00013588
2920
dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
YQECBLVSMFAWIZ-UHFFFAOYSA-N
dichloro(dimethyl)germane
73724
95+%
1529-48-2
Colorless
123°C
95% min
C2H6Cl2Ge
(CH3)2GeCl2
1 g
1.5
Solubility in water: reacts
C[Ge](C)(Cl)Cl
173.57
173.57
Liquid

Chemical Identifiers

1529-48-2
173.57
YQECBLVSMFAWIZ-UHFFFAOYSA-N
73724
C[Ge](C)(Cl)Cl
C2H6Cl2Ge
MFCD00013588
dimethylgermanium dichloride, germane, dichlorodimethyl, dimethyldichlorogermane, dichloro dimethyl germane, dimethylgermanedichloride, dimethylgermyl dichloride, geme2cl2, acmc-1boyh, germanium dimethyl dichloride, dimethyl germanium dichloride
dichloro(dimethyl)germane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 216-216-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only