Learn More
(Dichloromethylene)dimethylammonium chloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1230014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
(Dichloromethylene)dimethylammonium chloride | |
∼185°C (decomposition) | |
MFCD00011870 | |
UN3261 | |
Moisture sensitive | |
NRNFKRFWZQQDMD-UHFFFAOYSA-M | |
dichloromethylidene(dimethyl)azanium;chloride | |
2723945 | |
95% |
33842-02-3 | |
C3H6Cl3N | |
25 g | |
4157987 | |
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride | |
[Cl-].CN(C)[C+](Cl)Cl | |
162.44 | |
162.45 |
Chemical Identifiers
33842-02-3 | |
162.44 | |
NRNFKRFWZQQDMD-UHFFFAOYSA-M | |
2723945 | |
[Cl-].CN(C)[C+](Cl)Cl |
C3H6Cl3N | |
MFCD00011870 | |
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride | |
dichloromethylidene(dimethyl)azanium;chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 251-695-4
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only