missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Dichloromethylene)dimethylammonium chloride, 95%, Thermo Scientific Chemicals

$197.23 - $529.59

Chemical Identifiers

CAS 33842-02-3
Molecular Formula C3H6Cl3N
Molecular Weight (g/mol) 162.44
MDL Number MFCD00011870
InChI Key NRNFKRFWZQQDMD-UHFFFAOYSA-M
Synonym dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
PubChem CID 2723945
IUPAC Name dichloromethylidene(dimethyl)azanium;chloride
SMILES [Cl-].CN(C)[C+](Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1230014
View Documents
Thermo Scientific Chemicals
A1230014
25 g
Each for $197.23
 
AAA1230022
View Documents
Thermo Scientific Chemicals
A1230022
100 g
Each for $529.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33842-02-3
162.44
NRNFKRFWZQQDMD-UHFFFAOYSA-M
2723945
[Cl-].CN(C)[C+](Cl)Cl
C3H6Cl3N
MFCD00011870
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
dichloromethylidene(dimethyl)azanium;chloride

Specifications

33842-02-3
C3H6Cl3N
25 g
4157987
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
[Cl-].CN(C)[C+](Cl)Cl
162.44
162.45
(Dichloromethylene)dimethylammonium chloride
∼185°C (decomposition)
MFCD00011870
UN3261
Moisture sensitive
NRNFKRFWZQQDMD-UHFFFAOYSA-M
dichloromethylidene(dimethyl)azanium;chloride
2723945
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 251-695-4

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only