missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Dichloromethylene)dimethylammonium chloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1230022

 View more versions of this product

Catalog No. AAA1230022


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(Dichloromethylene)dimethylammonium chloride
∼185°C (decomposition)
MFCD00011870
UN3261
Moisture sensitive
NRNFKRFWZQQDMD-UHFFFAOYSA-M
dichloromethylidene(dimethyl)azanium;chloride
2723945
95%
33842-02-3
C3H6Cl3N
100 g
4157987
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
[Cl-].CN(C)[C+](Cl)Cl
162.44
162.45

Chemical Identifiers

33842-02-3
162.44
NRNFKRFWZQQDMD-UHFFFAOYSA-M
2723945
[Cl-].CN(C)[C+](Cl)Cl
C3H6Cl3N
MFCD00011870
dichloromethylene dimethylammonium chloride, phosgeniminium chloride, n-dichloromethylene-n-methylmethanaminium chloride, methanaminium, n-dichloromethylene-n-methyl-, chloride, dichloromethylene-dimethyliminium chloride, dichloromethylenedimethylammonium chloride, dichloromethylidene dimethylazanium chloride, phosgeneiminium chloride, phosgene-iminium chloride
dichloromethylidene(dimethyl)azanium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 251-695-4

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only